Learn More
Ace Glass, Inc. 2000mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, #11Ace-Thred inlet/outlets, 100mm 5 neck head, addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972923
Encompass
2000mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, #11Ace-Thred inlet/outlets, 100mm 5 neck head, addition funnel, condenser, pressure manifold and stirring assembly
Catalog No. 50-972-923 Supplier Ace Glass, Inc. Supplier No. 6429437
Add to Cart
Add to Cart
Request a Quote
2000mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, #11Ace-Thred inlet/outlets, 100mm 5 neck head, addition funnel, condenser, pressure manifold and stirring assembly