Learn More
Ace Glass, Inc. 2000mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, #11 Ace-Thred inlet/outlet, complete with addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50978724
Encompass
2000mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a #11 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.
Catalog No. 50-978-724 Supplier Ace Glass, Inc. Supplier No. 6438350
Add to Cart
Add to Cart
Request a Quote
2000mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a #11 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.