Learn More
Ace Glass, Inc. 3000mL, two-piece jacketed pressure reactor complete with0-10mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 150mm 6 neck head, addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972915
Encompass
3000mL, two-piece jacketed pressure reactor complete with0-10mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 150mm 6 neck head, addition funnel, condenser, pressure manifold and stirring assembly
Catalog No. 50-972-915 Supplier Ace Glass, Inc. Supplier No. 6429242
Add to Cart
Add to Cart
Request a Quote
3000mL, two-piece jacketed pressure reactor complete with0-10mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 150mm 6 neck head, addition funnel, condenser, pressure manifold and stirring assembly