Learn More
Ace Glass, Inc. 500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972899
Encompass
500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly
Catalog No. 50-972-899 Supplier Ace Glass, Inc. Supplier No. 6427407
Add to Cart
Add to Cart
Request a Quote
500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly