Learn More
Ace Glass, Inc. 500mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 60mm 4 neck head, addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972912
Encompass
500mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 60mm 4 neck head, addition funnel, condenser, pressure manifold and stirring assembly
Catalog No. 50-972-912 Supplier Ace Glass, Inc. Supplier No. 6429229
Add to Cart
Add to Cart
Request a Quote
500mL, two-piece jacketed pressure reactor complete with0-8mm valve and 28/15 ball joint bottom outlet, 28/15 ballpoint inlet/outlets, 60mm 4 neck head, addition funnel, condenser, pressure manifold and stirring assembly