Learn More
Ace Glass, Inc. 500mL, one-piece jacketed pressure reactor with #25 center, (3)#15, (1)#7 side necks, #11 Ace-Thred inlet/outlet, complete with addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972952
Encompass
500mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a #11 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.
Catalog No. 50-972-952 Supplier Ace Glass, Inc. Supplier No. 6438340
Add to Cart
Add to Cart
Request a Quote
500mL one-piece jacketed pressure reactor with a #25 center neck, (3) #15 and (1) #7 side necks, and a #11 Ace-Thred connection for 3/8 tubing on the jacket inlet/outlet. Reactor system designed to perform low to moderate positive pressure reactions, synthesis and catalysis. Rated at 45psi at 100 degrees Centigrade. Supplied with addition funnel, condenser, pressure manifold, and stirring assembly.