Learn More
Ace Glass, Inc. 500mL, two-piece jacketed pressure reactor, complete with28/15 ball joint inlet/outlets, 100mm head with #15 center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly
SDP
Catalog No. 50972886
Encompass
500mL, two-piece jacketed pressure reactor, complete with28/15 ball joint inlet/outlets, 100mm head with #15 center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly
Catalog No. 50-972-886 Supplier Ace Glass, Inc. Supplier No. 6427209
Add to Cart
Add to Cart
Request a Quote
500mL, two-piece jacketed pressure reactor, complete with28/15 ball joint inlet/outlets, 100mm head with #15 center, (3)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and stirring assembly