Learn More
Ace Glass, Inc. 500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and super magnet stirring assembly
SDP
Catalog No. 50972903
Encompass
500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and super magnet stirring assembly
Catalog No. 50-972-903 Supplier Ace Glass, Inc. Supplier No. 6427464
Add to Cart
Add to Cart
Request a Quote
500mL, two-piece jacketed pressure reactor, complete with#11 Ace-Thred inlet/outlets, 60mm head with #15 center, (2)#15, (1)#7 side necks, addition funnel, condenser, pressure manifold and super magnet stirring assembly