Learn More
Chemglass Life Sciences Chemglass Reaction Vessel Lid, 6-Neck, 50 mL (CG-MR-50L2) - Mettler EasyMax compatible
SDP
Catalog No. NC3073757
Encompass_Preferred
Chemglass CG-MR-50L2 is a six-neck reaction vessel lid for 50 mL laboratory reactors, designed for bench-scale synthesis and process development. The lid provides multiple sealed access ports for probes, additions, and fittings, enabling flexible setups and integration with automated reactor platforms.
  • Six-neck configuration for multiple fittings and probes.
  • 50 mL capacity suitable for small-scale reactions.
  • Joint sizes: (1) 14/20 standard neck with 10/18 outer; (2) two GL-14 side necks; (1) 7/15 neck; (1) 10/18 neck.
  • Borosilicate glass construction for chemical resistance and thermal stability.
  • Designed for use with automated bench-top reactor systems.
  • In-stock availability for immediate replacement.
Catalog No. NC3073757 Supplier Chemglass Life Sciences Supplier No. CGMR50L2
Chemglass CG-MR-50L2 is a six-neck reaction vessel lid for 50 mL laboratory reactors, designed for bench-scale synthesis and process development. The lid provides multiple sealed access ports for probes, additions, and fittings, enabling flexible setups and integration with automated reactor platforms.
  • Six-neck configuration for multiple fittings and probes.
  • 50 mL capacity suitable for small-scale reactions.
  • Joint sizes: (1) 14/20 standard neck with 10/18 outer; (2) two GL-14 side necks; (1) 7/15 neck; (1) 10/18 neck.
  • Borosilicate glass construction for chemical resistance and thermal stability.
  • Designed for use with automated bench-top reactor systems.
  • In-stock availability for immediate replacement.