Learn More
Chemglass Life Sciences Reaction Vessel Lid, 6-Nk, 50mL, Equivalent to Mettler Easymax Part # 51160817
SDP
Catalog No. 501532048
Encompass_Preferred
Reaction Vessel Lid, 6-Nk, 50mL, Compatible with Mettler Systems. (1) 14/20 SN with 10/18 Outer, (2) GL-14 SN's, (1) 7/12 SN and (1) 10/18 SN. Equivalent to Mettler Part # 51160817
Catalog No. 50-153-2048 Supplier Chemglass Life Sciences Supplier No. CGMR50L3
Reaction Vessel Lid, 6-Nk, 50mL, Compatible with Mettler Systems. (1) 14/20 SN with 10/18 Outer, (2) GL-14 SN's, (1) 7/12 SN and (1) 10/18 SN. Equivalent to Mettler Part # 51160817