Learn More
Chemglass Life Sciences ChemRxnHub TRIPLE Support Stand ONLY
SDP
Catalog No. 501531297
Encompass_Preferred
ChemRxnHub TRIPLE Support Stand ONLY. This unique stationary reactor stand is designed to fit into most benchtop hoods. ChemRxnHub Triple Stand can hold three reactors from 300mL to 5L. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, and Temperature Monitor. Stand includes removable spill tray. The weight of the stand is 128lbs.
Catalog No. 50-153-1297 Supplier Chemglass Life Sciences Supplier No. CG1949X30
Add to Cart
Add to Cart
Request a Quote
ChemRxnHub TRIPLE Support Stand ONLY. This unique stationary reactor stand is designed to fit into most benchtop hoods. ChemRxnHub Triple Stand can hold three reactors from 300mL to 5L. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, and Temperature Monitor. Stand includes removable spill tray. The weight of the stand is 128lbs.