Learn More
Chemglass Life Sciences ChemRxnHub DUAL Support Stand ONLY
SDP
Catalog No. 501531295
Encompass_Preferred
ChemRxnHub DUAL Support Stand ONLY. This unique stationary reactor stand is designed to fit into most benchtop hoods. ChemRxnHub Dual Stand can hold two reactors from 300mL to 5L. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, and Temperature Monitor. Stand includes removable spill tray. The weight of the stand is 68lbs.
Catalog No. 50-153-1295 Supplier Chemglass Life Sciences Supplier No. CG1949X20
Add to Cart
Add to Cart
Request a Quote
ChemRxnHub DUAL Support Stand ONLY. This unique stationary reactor stand is designed to fit into most benchtop hoods. ChemRxnHub Dual Stand can hold two reactors from 300mL to 5L. The stand provides support for the Reaction Vessel(s), Glassware, Overhead Stirring Motor, and Temperature Monitor. Stand includes removable spill tray. The weight of the stand is 68lbs.