Learn More
Chemglass Life Sciences Chemglass 10 L Process Reactor System, Complete, Electric Motor with Mechanical Bearing, 5-Neck Lid
SDP
Catalog No. NC3363898
Encompass_Preferred
The Chemglass 10 L process reactor is a complete, jacketed glass reaction system on an open-frame, auto-centering stand designed for pilot-scale synthesis and process development. It combines a 10 L jacketed vessel, 5-neck lid with powder addition funnel, integrated manifold for fluid handling, and an electric motor drive with separate digital controller for controlled stirring and temperature monitoring.
  • 10 L jacketed glass reactor vessel
  • Electric motor with separate digital controller
  • Mechanical seal stirrer bearing
  • Open-frame auto-center stand with heavy-duty lockable casters
  • 5-neck lid with large powder addition funnel
  • Zero dead space detachable drain valve
  • Manifold system for fluid transfers
  • Digital temperature monitor
  • Reactor jacket graduated on both sides
  • Support bushings for optional safety shield
Catalog No. NC3363898 Supplier Chemglass Life Sciences Supplier No. CG1965MB610M
Add to Cart
Add to Cart
Request a Quote
The Chemglass 10 L process reactor is a complete, jacketed glass reaction system on an open-frame, auto-centering stand designed for pilot-scale synthesis and process development. It combines a 10 L jacketed vessel, 5-neck lid with powder addition funnel, integrated manifold for fluid handling, and an electric motor drive with separate digital controller for controlled stirring and temperature monitoring.
  • 10 L jacketed glass reactor vessel
  • Electric motor with separate digital controller
  • Mechanical seal stirrer bearing
  • Open-frame auto-center stand with heavy-duty lockable casters
  • 5-neck lid with large powder addition funnel
  • Zero dead space detachable drain valve
  • Manifold system for fluid transfers
  • Digital temperature monitor
  • Reactor jacket graduated on both sides
  • Support bushings for optional safety shield