Learn More
Chemglass Life Sciences Chemglass 30 L Jacketed Process Reactor System, Complete, Electric Motor, Mechanical Bearing
SDP
Catalog No. NC3363900
Encompass_Preferred
The Chemglass 30 L jacketed process reactor system is a complete, open-frame reactor on an open-air stand designed for medium-scale synthesis and processing. It includes an electric motor with mechanical bearing, a multi-neck lid, an integrated manifold, and options for digital temperature monitoring and motor control.
  • 30 L capacity for medium-scale reactions.
  • Jacketed glass vessel with visible graduation scale on both sides.
  • Seven-neck lid with large powder addition port.
  • Zero-dead-space detachable drain valve with laser-etched directional arrows.
  • Mechanical seal stirrer bearing and motor mount arm that unlocks and swings out.
  • Open-air support frame with stainless steel uprights and lockable casters.
  • Includes manifold system for process connections.
  • Stainless steel inlet/outlet adapters available in M16 x 1 and M30 x 1.5.
  • Digital temperature monitor and motor controller options.
Catalog No. NC3363900 Supplier Chemglass Life Sciences Supplier No. CG1968MB630M
Add to Cart
Add to Cart
Request a Quote
The Chemglass 30 L jacketed process reactor system is a complete, open-frame reactor on an open-air stand designed for medium-scale synthesis and processing. It includes an electric motor with mechanical bearing, a multi-neck lid, an integrated manifold, and options for digital temperature monitoring and motor control.
  • 30 L capacity for medium-scale reactions.
  • Jacketed glass vessel with visible graduation scale on both sides.
  • Seven-neck lid with large powder addition port.
  • Zero-dead-space detachable drain valve with laser-etched directional arrows.
  • Mechanical seal stirrer bearing and motor mount arm that unlocks and swings out.
  • Open-air support frame with stainless steel uprights and lockable casters.
  • Includes manifold system for process connections.
  • Stainless steel inlet/outlet adapters available in M16 x 1 and M30 x 1.5.
  • Digital temperature monitor and motor controller options.