Learn More
Chemglass Life Sciences Chemglass 50 L Jacketed Process Reactor System, Complete, Air Motor
SDP
Catalog No. NC3363899
Encompass_Preferred
Complete 50 L jacketed process reactor system with air-driven motor, multi-neck lid, digital temperature monitoring, and a compact stainless-steel support frame with lockable casters. Designed for process mixing, heating and cooling through the reactor jacket, it provides easy access for loading and maintenance.
  • Jacketed 50 L borosilicate glass reaction vessel.
  • Air-driven motor with swing-out motor mount for improved access.
  • Digital temperature monitor for process control.
  • Seven-neck lid with large powder addition port.
  • Zero-dead-space detachable drain valve for efficient transfer.
  • Compact stainless steel support frame with lockable casters.
  • PTFE-coated aluminum fittings and stainless steel inlet/outlet adapters.
  • Graduated jacket scale and lower safety ring for level monitoring.
Catalog No. NC3363899 Supplier Chemglass Life Sciences Supplier No. CG1970650M
Add to Cart
Add to Cart
Request a Quote
Complete 50 L jacketed process reactor system with air-driven motor, multi-neck lid, digital temperature monitoring, and a compact stainless-steel support frame with lockable casters. Designed for process mixing, heating and cooling through the reactor jacket, it provides easy access for loading and maintenance.
  • Jacketed 50 L borosilicate glass reaction vessel.
  • Air-driven motor with swing-out motor mount for improved access.
  • Digital temperature monitor for process control.
  • Seven-neck lid with large powder addition port.
  • Zero-dead-space detachable drain valve for efficient transfer.
  • Compact stainless steel support frame with lockable casters.
  • PTFE-coated aluminum fittings and stainless steel inlet/outlet adapters.
  • Graduated jacket scale and lower safety ring for level monitoring.