Learn More
Chemglass Life Sciences Chemglass 50 L Squatty Jacketed Glass Process Reactor System, 400 mm Flange, Electric Motor (CG-1968-MB651M)
SDP
Catalog No. NC3593844
Encompass_Preferred
A 50 L squat, jacketed, cylindrical glass reactor system designed for pilot-scale processing. The low-profile vessel and integrated brushless DC overhead drive provide controlled mixing and efficient heat transfer, while the heavy-duty support frame and casters enable mobility and easy access for cleaning and draining.
  • 50 L jacketed squat cylindrical glass vessel
  • 400 mm flat top flange for large access and connections
  • Brushless DC overhead stirrer with variable speed (~30-500 rpm)
  • Heavy-duty stainless steel support frame with mobile swivel casters
  • Maximized clearance below the drain valve for easy draining
  • Jacketed design for temperature control and heat transfer
Catalog No. NC3593844 Supplier Chemglass Life Sciences Supplier No. CG1968MB651M
Add to Cart
Add to Cart
Request a Quote
A 50 L squat, jacketed, cylindrical glass reactor system designed for pilot-scale processing. The low-profile vessel and integrated brushless DC overhead drive provide controlled mixing and efficient heat transfer, while the heavy-duty support frame and casters enable mobility and easy access for cleaning and draining.
  • 50 L jacketed squat cylindrical glass vessel
  • 400 mm flat top flange for large access and connections
  • Brushless DC overhead stirrer with variable speed (~30-500 rpm)
  • Heavy-duty stainless steel support frame with mobile swivel casters
  • Maximized clearance below the drain valve for easy draining
  • Jacketed design for temperature control and heat transfer