Learn More
Enterprise Technology Solutions Read/Right Duster, Dust-Free, 5" Extension Wand, 10 oz.
SDP
Catalog No. 5000740574
Encompass_Preferred
All-purpose, economical gas duster safely removes lint and dust from keyboards, computers and other office machines. Formula is 100 percent ozone-safe. Duster is equipped with an extension wand to get to hard-to-reach places. Duster also contains bitterant to discourage inhalant abuse.
Catalog No. 50-007-40574 Supplier Enterprise Technology Solutions Supplier No. REARR3700
All-purpose, economical gas duster safely removes lint and dust from keyboards, computers and other office machines. Formula is 100 percent ozone-safe. Duster is equipped with an extension wand to get to hard-to-reach places. Duster also contains bitterant to discourage inhalant abuse.