Learn More
Enterprise Technology Solutions Read/Right Magnetic Head Cleaner, 2-1/2"x5-1/4"x4-1/2", 25/BX
SDP
Catalog No. 5000740280
Encompass_Preferred
CardKleen optimizes the performance of the magnetic strip on point-of-sales card readers. Cleaner is designed to clean the magnetic heads quickly and safely on point-of-sale credit card readers, access control/ID readers, and all open systems. Presaturated, individually packed cards are nonabrasive and easy to use.
Catalog No. 50-007-40280 Supplier Enterprise Technology Solutions Supplier No. REARR1222
CardKleen optimizes the performance of the magnetic strip on point-of-sales card readers. Cleaner is designed to clean the magnetic heads quickly and safely on point-of-sale credit card readers, access control/ID readers, and all open systems. Presaturated, individually packed cards are nonabrasive and easy to use.