Learn More
Enterprise Technology Solutions Read/Right One Step Screen Cleaning Wipes, 100/BX
SDP
Catalog No. 5000739648
Encompass_Preferred
Ultra-thin, high-quality wipes are presaturated with special nonstreaking cleaning and antistatic solution that works quickly, evaporating in seconds. Use on microcomputer CRT screens, VDT and TV displays, instrument glass and micrographics screens. Towelettes are not for use on nylon mesh or polarized antiglare screens. Towelettes are individually wrapped in foil packets.
Catalog No. 50-007-39648 Supplier Enterprise Technology Solutions Supplier No. REARR1309
Ultra-thin, high-quality wipes are presaturated with special nonstreaking cleaning and antistatic solution that works quickly, evaporating in seconds. Use on microcomputer CRT screens, VDT and TV displays, instrument glass and micrographics screens. Towelettes are not for use on nylon mesh or polarized antiglare screens. Towelettes are individually wrapped in foil packets.