Learn More
Enterprise Technology Solutions Read/Right Screen Cleaning Pads, Twin Packs, Wet/Dry Wipes, 14 Sets/BX
SDP
Catalog No. 5000740173
Encompass_Preferred
Kleen and Dry Screen Cleaning Pads offer a highly efficient, two-part method for cleaning and controlling static on terminal and display screens. One packet contains a pre-moistened pad with an exclusive antistatic cleaner solution that controls dangerous static charges while removing vision-inhibiting contaminants from the screen surface. The other packet contains a dry absorbent, lint-free cloth for wiping the screen dry and clear. Pads leave screen optically clear and lint-free. Pads are not for use on nylon mesh or polarized antiglare filters.
Catalog No. 50-007-40173 Supplier Enterprise Technology Solutions Supplier No. REARR1205
Kleen and Dry Screen Cleaning Pads offer a highly efficient, two-part method for cleaning and controlling static on terminal and display screens. One packet contains a pre-moistened pad with an exclusive antistatic cleaner solution that controls dangerous static charges while removing vision-inhibiting contaminants from the screen surface. The other packet contains a dry absorbent, lint-free cloth for wiping the screen dry and clear. Pads leave screen optically clear and lint-free. Pads are not for use on nylon mesh or polarized antiglare filters.