Learn More
Enterprise Technology Solutions Read/Right Screen Cleaning Wipes,Presaturated,Nonstreaking,24/BX,WE
SDP
Catalog No. 5000740174
Encompass_Preferred
Ultra-thin, screen-cleaning wipes are presaturated with special nonstreaking, antistatic solution that works quickly and evaporates in seconds. Use on microcomputer CRT screens, VDT and TV displays, instrument glass and micrographics screens. Not for use on nylon mesh/polarized antiglare screens. Towelettes are individually wrapped in foil packets.
Catalog No. 50-007-40174 Supplier Enterprise Technology Solutions Supplier No. REARR1209
Ultra-thin, screen-cleaning wipes are presaturated with special nonstreaking, antistatic solution that works quickly and evaporates in seconds. Use on microcomputer CRT screens, VDT and TV displays, instrument glass and micrographics screens. Not for use on nylon mesh/polarized antiglare screens. Towelettes are individually wrapped in foil packets.