Learn More
Molecular Devices Molecular Devices Microplate Washer, 96-Well Wash Head, Two Reagent Ports
Catalog No. 5000014361
Encompass_Preferred
The Molecular Devices AquaMax 2000 Microplate Washer (configured with a 96-well wash head) is a compact, self-contained benchtop washer that delivers reproducible, high-precision washing for biochemical and cell-based assays. It offers interchangeable plate and cell wash heads, two reagent ports, and seamless integration with common automation and stacker systems.
  • Interchangeable 96- and 384-well wash heads for flexible assay formats.
  • Two reagent ports to support multiple wash protocols without bottle changes.
  • Supports 4 L and 10 L reagent and waste reservoirs for extended runs.
  • Dispense volume range 20-400 μL for accurate well washing.
  • Compact footprint and touchscreen controls for easy benchtop use.
Catalog No. 50-000-14361 Supplier Molecular Devices Supplier No. AQUAMAX2KBUNDLE
Add to Cart
Add to Cart
Request a Quote
The Molecular Devices AquaMax 2000 Microplate Washer (configured with a 96-well wash head) is a compact, self-contained benchtop washer that delivers reproducible, high-precision washing for biochemical and cell-based assays. It offers interchangeable plate and cell wash heads, two reagent ports, and seamless integration with common automation and stacker systems.
  • Interchangeable 96- and 384-well wash heads for flexible assay formats.
  • Two reagent ports to support multiple wash protocols without bottle changes.
  • Supports 4 L and 10 L reagent and waste reservoirs for extended runs.
  • Dispense volume range 20-400 μL for accurate well washing.
  • Compact footprint and touchscreen controls for easy benchtop use.