Learn More
Molecular Devices Molecular Devices Microplate Reader, Multimode, 200-1000 nm, 6-384 Wells
Catalog No. 5000014683
Encompass_Preferred
The SpectraMax M2 is a dual-monochromator multimode microplate reader for absorbance, fluorescence, time-resolved fluorescence, and luminescence assays across 6-384 well plates and cuvette formats.
  • Multimode detection for absorbance, fluorescence intensity, time-resolved fluorescence, and luminescence.
  • Wavelength range: absorbance 200-1000 nm, fluorescence excitation 250-850 nm, fluorescence emission 360-850 nm.
  • Supports 6, 12, 24, 48, 96, and 384 well plate formats.
  • Dual-mode cuvette port for cuvette-based assays and dual monochromators for tunable wavelength selection.
  • Power: 100-240 Vac, 3 A, 50/60 Hz; power consumption under 125 W.
  • Includes PathCheck optical pathlength normalization, kinetic and endpoint read modes, and robot compatibility.
Catalog No. 50-000-14683 Supplier Molecular Devices Supplier No. M2G
Add to Cart
Add to Cart
Request a Quote
The SpectraMax M2 is a dual-monochromator multimode microplate reader for absorbance, fluorescence, time-resolved fluorescence, and luminescence assays across 6-384 well plates and cuvette formats.
  • Multimode detection for absorbance, fluorescence intensity, time-resolved fluorescence, and luminescence.
  • Wavelength range: absorbance 200-1000 nm, fluorescence excitation 250-850 nm, fluorescence emission 360-850 nm.
  • Supports 6, 12, 24, 48, 96, and 384 well plate formats.
  • Dual-mode cuvette port for cuvette-based assays and dual monochromators for tunable wavelength selection.
  • Power: 100-240 Vac, 3 A, 50/60 Hz; power consumption under 125 W.
  • Includes PathCheck optical pathlength normalization, kinetic and endpoint read modes, and robot compatibility.