Learn More
Molecular Devices Molecular Devices Spectrophotometer, Micro-Volume Port, Cuvette Port, 190-1100 nm
Catalog No. 5000014679
Encompass_Preferred
The SpectraMax QuickDrop Micro-Volume Spectrophotometer provides rapid, low-volume UV-Vis measurements for DNA, RNA, and protein quantitation using both a micro-volume sample port and a standard cuvette port.
  • Micro-volume sample port with minimum volume 0.5 μL.
  • Micro-volume pathlength 0.5 mm for low-volume measurements.
  • Cuvette port with pathlength options of 1 mm, 2 mm, 5 mm, and 10 mm.
  • Full-spectrum wavelength range 190-1100 nm for broad UV-Vis analysis.
  • Pulsed xenon lamp and 1024-element CCD detector for rapid, full-spectrum acquisition.
  • 7 in color capacitive touchscreen for instrument control and data review.
  • Absorbance range suitable for low to high concentration measurements.
  • Compact footprint, approximately 190 x 290 x 410 mm and weight under 4 kg.
Catalog No. 50-000-14679 Supplier Molecular Devices Supplier No. QUICKDROP
Add to Cart
Add to Cart
Request a Quote
The SpectraMax QuickDrop Micro-Volume Spectrophotometer provides rapid, low-volume UV-Vis measurements for DNA, RNA, and protein quantitation using both a micro-volume sample port and a standard cuvette port.
  • Micro-volume sample port with minimum volume 0.5 μL.
  • Micro-volume pathlength 0.5 mm for low-volume measurements.
  • Cuvette port with pathlength options of 1 mm, 2 mm, 5 mm, and 10 mm.
  • Full-spectrum wavelength range 190-1100 nm for broad UV-Vis analysis.
  • Pulsed xenon lamp and 1024-element CCD detector for rapid, full-spectrum acquisition.
  • 7 in color capacitive touchscreen for instrument control and data review.
  • Absorbance range suitable for low to high concentration measurements.
  • Compact footprint, approximately 190 x 290 x 410 mm and weight under 4 kg.