Learn More
Molecular Devices Molecular Devices Multi-Mode Microplate Reader, Absorbance, Fluorescence, Luminescence
Catalog No. 5000014466
Encompass_Preferred
The SpectraMax Mini Multi-Mode Microplate Reader is a compact, user-upgradeable instrument providing absorbance, fluorescence, and luminescence detection with integrated SoftMax Pro data analysis. It supports 6-384 well plates and temperature control for routine and kinetic assays.
  • Compact, user-upgradeable design with two- or three-mode configurations.
  • Supports 6, 12, 24, 48, 96, and 384-well plates.
  • Detects absorbance (200-1000 nm), fluorescence (250-850 nm), and luminescence.
  • Offers endpoint, kinetic, spectral, and well scanning read modes.
  • Temperature control from ambient +4°C to 45°C with ±1°C uniformity at 37°C.
  • Includes advanced data analysis software and auto-adjusting PMT for wide dynamic range.
Catalog No. 50-000-14466 Supplier Molecular Devices Supplier No. SMAXMINIAF
Add to Cart
Add to Cart
Request a Quote
The SpectraMax Mini Multi-Mode Microplate Reader is a compact, user-upgradeable instrument providing absorbance, fluorescence, and luminescence detection with integrated SoftMax Pro data analysis. It supports 6-384 well plates and temperature control for routine and kinetic assays.
  • Compact, user-upgradeable design with two- or three-mode configurations.
  • Supports 6, 12, 24, 48, 96, and 384-well plates.
  • Detects absorbance (200-1000 nm), fluorescence (250-850 nm), and luminescence.
  • Offers endpoint, kinetic, spectral, and well scanning read modes.
  • Temperature control from ambient +4°C to 45°C with ±1°C uniformity at 37°C.
  • Includes advanced data analysis software and auto-adjusting PMT for wide dynamic range.