Learn More
Aat Bioquest ReadiLink™ KLH Conjugation Kit, 2 Reactions
SDP
Catalog No. NC3791297
Encompass_Preferred
Keyhole Limpet Hemocyanin (KLH) is one of the most commonly used carriers in the conjugation of peptides for antibody production. Mariculture keyhole limpet hemocyanin (mcKLH) is a hemocyanin from the Concholepas concholepas mollusk with immunogenic properties similar to KLH but is more stable and efficient as a carrier protein for the production of antibodies to haptens and peptides. It contains numerous sites per molecule for effective conjugation of peptides and other antigens using amine-reactive or carboxyl-reactive crosslinkers. mcKLH is currently the industry standard for antibody production against a hapten or peptide. This ReadiLink™ KLH Conjugation kit is primarily optimized for the simple preparation of hapten-carrier conjugates for immunization and antibody production. The ReadiLink™ KLH Conjugation kit is one-step conjugation of a hapten to a carrier protein using the carboxyl-reactive carbodiimide as the crosslinker.
Catalog No. NC3791297 Supplier Aat Bioquest Supplier No. 5502
Keyhole Limpet Hemocyanin (KLH) is one of the most commonly used carriers in the conjugation of peptides for antibody production. Mariculture keyhole limpet hemocyanin (mcKLH) is a hemocyanin from the Concholepas concholepas mollusk with immunogenic properties similar to KLH but is more stable and efficient as a carrier protein for the production of antibodies to haptens and peptides. It contains numerous sites per molecule for effective conjugation of peptides and other antigens using amine-reactive or carboxyl-reactive crosslinkers. mcKLH is currently the industry standard for antibody production against a hapten or peptide. This ReadiLink™ KLH Conjugation kit is primarily optimized for the simple preparation of hapten-carrier conjugates for immunization and antibody production. The ReadiLink™ KLH Conjugation kit is one-step conjugation of a hapten to a carrier protein using the carboxyl-reactive carbodiimide as the crosslinker.