Learn More
Aat Bioquest ReadiLink Rapid iFluor 633 A
SDP
Catalog No. 502979148
Encompass_Preferred
ReadiLink Rapid iFluor 633 Antibody Labeling Kit *Microscale Optimized for Labeling 50 μg Antibody Per Reaction*
Catalog No. 50-297-9148 Supplier Aat Bioquest Supplier No. 1260
ReadiLink Rapid iFluor 633 Antibody Labeling Kit *Microscale Optimized for Labeling 50 μg Antibody Per Reaction*