Learn More
Aat Bioquest ReadiLink Rapid mFluor Viole
SDP
Catalog No. 502979352
Encompass_Preferred
ReadiLink Rapid mFluor Violet 420 Antibody Labeling Kit *Microscale Optimized for Labeling 50 μg Antibody Per Reaction*
Catalog No. 50-297-9352 Supplier Aat Bioquest Supplier No. 1105
ReadiLink Rapid mFluor Violet 420 Antibody Labeling Kit *Microscale Optimized for Labeling 50 μg Antibody Per Reaction*