Learn More
Aat Bioquest READILINK VIOLET 510 AB
SDP
Catalog No. NC2590249
Encompass_Preferred
AAT Bioquests mFluor dyes are developed for flow cytometry-focused applications. These dyes have large Stokes Shifts and can be well excited by the laser lines of flow cytometers. mFluor Violet 510 dyes have fluorescence excitation and emission maxima of 405 nm and 510 nm respectively. mFluor Violet 510 SE is reasonably stable and shows good reactivity and selectivity with protein amino groups. This kit has all the essential components to perform two separate labeling reactions with no column purification needed.
Catalog No. NC2590249 Supplier Aat Bioquest Supplier No. 1110
AAT Bioquests mFluor dyes are developed for flow cytometry-focused applications. These dyes have large Stokes Shifts and can be well excited by the laser lines of flow cytometers. mFluor Violet 510 dyes have fluorescence excitation and emission maxima of 405 nm and 510 nm respectively. mFluor Violet 510 SE is reasonably stable and shows good reactivity and selectivity with protein amino groups. This kit has all the essential components to perform two separate labeling reactions with no column purification needed.