Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Polystyrene ReadyCal Standards Kit, 4 mL, Calibration

Catalog No. 50785846 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Polystyrene ReadyCal Standards Kit provides a reliable and convenient solution for multi-point calibration curves, eliminating the need for manual weighing. It includes thirty autosampler vials, each with four polystyrene standards across three distinct molecular weight ranges. The kit offers a simple reconstitution process: add solvent, allow to stand, gently shake, and load into an autosampler. Detailed instructions are included.

  • Thirty autosampler vials, each with four polystyrene standards.
  • Three distinct molecular weight ranges (10 units each).
  • Enables quick, reliable multi-point calibration.
  • Simple reconstitution: add solvent, stand 2 hr, gently shake, load.
  • Designed for GPC/GFC/SEC separation; UV, RI/ELSD detection.
  • Solid formulation; store at 4°C (Fridge).
  • Shelf life: 36 months; 1 week reconstituted.
  • Shipping: Ambient.

Catalog No. 50-785-846 Supplier Waters Corp Supplier No. WAT058930
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Polystyrene ReadyCal Standards Kit provides a reliable and convenient solution for multi-point calibration curves, eliminating the need for manual weighing. It includes thirty autosampler vials, each with four polystyrene standards across three distinct molecular weight ranges. The kit offers a simple reconstitution process: add solvent, allow to stand, gently shake, and load into an autosampler. Detailed instructions are included.

  • Thirty autosampler vials, each with four polystyrene standards.
  • Three distinct molecular weight ranges (10 units each).
  • Enables quick, reliable multi-point calibration.
  • Simple reconstitution: add solvent, stand 2 hr, gently shake, load.
  • Designed for GPC/GFC/SEC separation; UV, RI/ELSD detection.
  • Solid formulation; store at 4°C (Fridge).
  • Shelf life: 36 months; 1 week reconstituted.
  • Shipping: Ambient.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.