Learn More
Waters Corp Waters Polystyrene ReadyCal Standard Kit, 2 mL, Multi-point Calibration, GPC/GFC/SEC
Catalog No. 50785847 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit contains thirty autosampler vials, each with four polystyrene standards, offering three distinct molecular weight ranges. It enables rapid and accurate generation of a multi-point calibration curve without weighing chemicals. Each vial contains a polymer mixture for baseline resolution of components.
  • Enables rapid and accurate generation of multi-point calibration curves without weighing chemicals.
  • Provided in autosampler vials, simplifying preparation and analysis.
  • Includes three distinct molecular weight ranges with 10 units of each.
  • Easy to use: simply add solvent, wait, shake, and load.
  • Comes with detailed instructions for proper usage.
  • Designed to pour solvent directly into the vial and mix.
Catalog No. 50-785-847 Supplier Waters Corp Supplier No. WAT058931
This kit contains thirty autosampler vials, each with four polystyrene standards, offering three distinct molecular weight ranges. It enables rapid and accurate generation of a multi-point calibration curve without weighing chemicals. Each vial contains a polymer mixture for baseline resolution of components.
  • Enables rapid and accurate generation of multi-point calibration curves without weighing chemicals.
  • Provided in autosampler vials, simplifying preparation and analysis.
  • Includes three distinct molecular weight ranges with 10 units of each.
  • Easy to use: simply add solvent, wait, shake, and load.
  • Comes with detailed instructions for proper usage.
  • Designed to pour solvent directly into the vial and mix.