Learn More
LGC GENOMICS LLC (Lucigen) Ready-Lyse Lysozyme Solution, 4,000,000 UReady-to-use lysozyme solution to be used with our nucleic acid purification kits.
Catalog No. NC9902425
Encompass
Ready-Lyse™ Lysozyme Solution is a recombinant lysozyme preparation, derived from a nonmammalian, nonavian source, that is useful for the lysis of Gram-negative (such as E. coli) and Gram-positive (such as Bacillus sp.) bacteria.
This item is not returnable. View return policy
Catalog No. NC9902425 Supplier LGC GENOMICS LLC Supplier No. R1804M
Ready-Lyse™ Lysozyme Solution is a recombinant lysozyme preparation, derived from a nonmammalian, nonavian source, that is useful for the lysis of Gram-negative (such as E. coli) and Gram-positive (such as Bacillus sp.) bacteria.