Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ready-Red™, MP Biomedicals™

Catalog No. MP1RYRED001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1RYRED001 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1RYRED001 Supplier MP Biomedicals, Inc Supplier No. 11RYRED001
Add to Cart
Add to Cart

Ready-Red™ Molecular biology grade, >99%, Liquid

  • Ready-Red™ is used either to remove traces of phenol from the aqueous phase or can be mixed directly with AquaPhenol™ (void volume in AquaPhenol™ bottle is adapted to this purpose).
  • Ready-Red™ is a coloured mixture of chloroform and isoamyl alcohol.
  • Ready-Red™ is a Chloroform:isoamyl alcohol 24:1 with red dye solution which facilitates separation of organic and aqueous phases. It improves recovery.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula CHCl3, C5H12O
Synonym Phenol removal reagent

Specifications

Physical Form Liquid
Quantity 250 mL
UN Number UN1888
Grade >99%
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.