Learn More
Alkali Scientific Ready Strain Cell Straining Kits™, Includes Strainer, 50mL Tube, Screw-cap, 70µm, White, Sterile, Individually Wrapped, 50/cs
SDP
Catalog No. 502388772
Encompass_Preferred
ReadyStrain is the first product of its kind, providing all the necessary parts for filtration of cell suspensions and combining them into a single, preassembled, ready to use, sterile packet. These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain packet directly under the hood. Each kit contains a sterile SureStrain Strainer, pre-attached to a sterile 50mL tube, with a separate sterile screw-cap ready for assembly, processing and storage after the filtration is complete. Kits are supplied in easy open “tear-strip” bags and are gamma sterilized.
Catalog No. 50-238-8772 Supplier Alkali Scientific Supplier No. MT5070
ReadyStrain is the first product of its kind, providing all the necessary parts for filtration of cell suspensions and combining them into a single, preassembled, ready to use, sterile packet. These kits eliminate the contamination risks associated with opening multiple packages, and assembling separate pieces (i.e. tubes strainers and caps). The individual sterile packets mean researchers are no longer forced to expose entire bags of 50mL tubes and corresponding caps in order to perform one filtration. Instead they can open a single ReadyStrain packet directly under the hood. Each kit contains a sterile SureStrain Strainer, pre-attached to a sterile 50mL tube, with a separate sterile screw-cap ready for assembly, processing and storage after the filtration is complete. Kits are supplied in easy open “tear-strip” bags and are gamma sterilized.