Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Ready-to-Load DNA Sequencing Kit
SDPSCI_ED

Catalog No. s29011 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S29011 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S29011 Supplier Edvotek™ Supplier No. 120
Only null left
Add to Cart
Add to Cart

Requires

Electrophoresis apparatus, power supply, automatic micropipette and tips, balance, microwave or hot plate, visualization system.

Used to introduce genome concepts and help students gain a better understanding of the science behind DNA sequencing. Edvotek™ Ready-to-Load DNA Sequencing Kit is for 8 lab groups.

Introduce your students to the exciting science of DNA Sequencing. This kit contains the four Ready-to-Load sequenced DNAs (nucleotides A, C, G, & T) in an easy to use, safe format. Students load the four separate reactions into agarose gels, run the gels, stain them, and actually read the DNA sequence. This experiment can be used to introduce genome concepts and help your students gain a better understanding of the science behind DNA sequencing.

Specifications

Grade 9 to Undergraduate
Includes Instructions, Ready-to-Load™ QuickStrip™ DNA samples, UltraSpec-Agarose™ powder, practice gel loading solution, electrophoresis buffer, InstaStain™ Blue and FlashBlue™ stain, calibrated pipette, and microtipped transfer pipettes
Time Range Complete in 60 minutes
Content And Storage Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.
Class Size 8 Lab Groups
Product Type Ready-to-Load DNA Sequencing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.