Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Ready-To-Use Lysing Bead Tubes for Bead Mill Homogenizers

Catalog No. 01922199 Shop All OHAUS Products
Change view
Click to view available options
Closure Color:
Blue
Brown
Green
Orange
Red
White
Yellow
Sample Type:
Animal Tissue
Bacteria
Environmental
Fungi
Plant
Soft Sample
Yeast
Particle Size:
1 μm
100 μm
100 μm, 800 μm, 3.0 mm
2.8 mm
3 μm
400 μm
800 μm
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Sample Type Particle Size
01-922-199 Blue Fungi 800 μm
01-922-203 Brown Environmental 100 μm, 800 μm, 3.0 mm
01-922-201 Green Plant 3 μm
01-922-200 Orange Soft Sample 1 μm
01-922-202 Red Animal Tissue 2.8 mm
01-922-197 White Bacteria 100 μm
01-922-198 Yellow Yeast 400 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 01-922-199 Supplier OHAUS™ Supplier No. 30391405
Add to Cart
Add to Cart

Protect the integrity of your samples - tubes, grinding beads and balls are treated and tested to be DNase and RNase-free.

  • Convenient wet or dry grinding
  • Seven distinct media types for plant, soft sample, fungi, yeast, bacteria or environmental sample processing
  • Threaded caps are color coded for easy identification
  • Applications:
    • DNA, RNA and Protein Analysis (from mammalian tissue, plant, soil, seed, fungi, bacteria, virus and other cells), Cell Lysing, PCR, Gene Expression
  • Design Features:
    • Tubes and grinding media are DNase/RNase-free
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lysing Bead Tubes
Capacity (Metric) 2 mL
Particle Size 800 μm
Material Polyethylene
Bottom Type Skirted Tube
Closure Color Blue
For Use With (Equipment) Homogenizers
Packaging Quantity 100 Pk.
Sample Type Fungi
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.