Learn More
Research Products International Corp Ready Towel Laboratory Towel Dispenser
SDP
Catalog No. 501367632
Encompass
ReadyTowel dispenser provides a convenient and quick access to a paper towel and preserves integrity to clean towels. Feature a unique and patented dispensing system that serves up the leading edge of a single towel each and every time. Ideal for laboratory use as fingers touch only the top towel and reduces the risk of contamination. ReadyTowel dispenser saves time and money by eliminating wasted towels. Accommodates all Multifold, A-Fold, M-Fold, C-Fold and Scottfold M paper towels. Dispenser housing is constructed of durable white acrylic with a clear PETG dispensing panel. Designed for placing on a benchtop or can be wall mounted with included hardware.
This item is not returnable. View return policy
Catalog No. 50-136-7632 Supplier Research Products International Corp Supplier No. 145642
ReadyTowel dispenser provides a convenient and quick access to a paper towel and preserves integrity to clean towels. Feature a unique and patented dispensing system that serves up the leading edge of a single towel each and every time. Ideal for laboratory use as fingers touch only the top towel and reduces the risk of contamination. ReadyTowel dispenser saves time and money by eliminating wasted towels. Accommodates all Multifold, A-Fold, M-Fold, C-Fold and Scottfold M paper towels. Dispenser housing is constructed of durable white acrylic with a clear PETG dispensing panel. Designed for placing on a benchtop or can be wall mounted with included hardware.