Learn More
Med Vet International Ready Warm Wall-mounted Fluid Bag Warmer, J0900R4TWS, Each
Catalog No. 5000583929
Encompass_Preferred
  The Ready Warm is a multi-use digital warming device for clinics, laboratories, and research facilities. It is available in 3 models: Wall-mounted Fluid Bag Warmer – for warming fluid bags (4 one-liter bags) while saving valuable bench space. • Digital • Wall mount CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: Ambient to 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz ADVANTAGES Compact design Ready Warm Wall-mounted Fluid Bag Warmer Actual Weight: 13 lb. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.  
Catalog No. 50-005-83929 Supplier Med Vet International Supplier No. JOR900R4TWS
  The Ready Warm is a multi-use digital warming device for clinics, laboratories, and research facilities. It is available in 3 models: Wall-mounted Fluid Bag Warmer – for warming fluid bags (4 one-liter bags) while saving valuable bench space. • Digital • Wall mount CONSTRUCTION Corrosion resistant aluminum Durable metal construction SPECIFICATIONS Temperature range: Ambient to 122°F +/-5°F Preset at 100°F (37°C) Accuracy: +/- 1.8°F WEIGHT AND DIMS Length: 22.835" (580mm) Width: 12" (305mm) Height: 3.07" (78mm) Weight: 13lbs (5.89kg) POWER / CONTROLS Lighted power switch Auto-switching CE / cUL approved power adapter Output: 12vDC / 10A Input: 100-240vAC 50/60Hz ADVANTAGES Compact design Ready Warm Wall-mounted Fluid Bag Warmer Actual Weight: 13 lb. Weight reflected in additional information is in reference to dimensional weight for shipping purposes.  