Learn More
Research Products International Corp Readyclean Tubes, Graduated, 1.5ml, Natural, 250/cs
SDP
Catalog No. 501978614
Encompass
Graduations marked at every 100μl to provide easy sample estimation. Solidly constructed for high speed centrifugation. Centrifugation Rating for all tubes is >20,000xg. Centrifuge Max for all tubes is >35,000xg (not routinely tested at this force). Certified RNase, DNase & DNA free. Sterile versions are certified free of pyrogens. All tubes are autoclavable. Maximum Recommended Temperature for all tubes is 121ºC. Minimum Recommended Temperature for all tubes is -90ºC. All tubes have a writing area.
Catalog No. 50-197-8614 Supplier Research Products International Corp Supplier No. 145515S
Graduations marked at every 100μl to provide easy sample estimation. Solidly constructed for high speed centrifugation. Centrifugation Rating for all tubes is >20,000xg. Centrifuge Max for all tubes is >35,000xg (not routinely tested at this force). Certified RNase, DNase & DNA free. Sterile versions are certified free of pyrogens. All tubes are autoclavable. Maximum Recommended Temperature for all tubes is 121ºC. Minimum Recommended Temperature for all tubes is -90ºC. All tubes have a writing area.