Learn More
Gold Biotechnology Inc ReadyLadder 1kb PLS DNA Ladder
SDP
Catalog No. 5000562164
Encompass_Preferred
GoldBio s ReadyLadder 1 kb PLUS DNA Ladder is a reliable ready-to-use molecular weight marker designed for accurate sizing of medium-to-large DNA fragments Composed of 14 distinct bands ranging from 250 bp to 25 kb this ladder provides excellent coverage for routine electrophoresis
Catalog No. 50-005-62164 Supplier Gold Biotechnology Inc Supplier No. D0111000
GoldBio s ReadyLadder 1 kb PLUS DNA Ladder is a reliable ready-to-use molecular weight marker designed for accurate sizing of medium-to-large DNA fragments Composed of 14 distinct bands ranging from 250 bp to 25 kb this ladder provides excellent coverage for routine electrophoresis