Learn More
Gold Biotechnology Inc ReadyLadderTM 1 kb DNA Ladder
SDP
Catalog No. 5000562040
Encompass_Preferred
The ReadyLadder 1 kb DNA Ladder from GoldBio offers a convenient ready-to-use solution for accurate DNA sizing in agarose gel electrophoresis Featuring 13 defined bands spanning 250 bp to 10 kb it provides the resolution needed for routine molecular biology experiments
Catalog No. 50-005-62040 Supplier Gold Biotechnology Inc Supplier No. D0101500
The ReadyLadder 1 kb DNA Ladder from GoldBio offers a convenient ready-to-use solution for accurate DNA sizing in agarose gel electrophoresis Featuring 13 defined bands spanning 250 bp to 10 kb it provides the resolution needed for routine molecular biology experiments