Learn More
Gold Biotechnology Inc ReadyLadderTM 1 kb DNA Ladder
Supplier: Gold Biotechnology Inc D0103000
The ReadyLadder 1 kb DNA Ladder from GoldBio offers a convenient ready-to-use solution for accurate DNA sizing in agarose gel electrophoresis Featuring 13 defined bands spanning 250 bp to 10 kb it provides the resolution needed for routine molecular biology experiments