Learn More
Gold Biotechnology Inc ReadyLadderTM 100 bp PLUSTM
SDP
Catalog No. 5000562121
Encompass_Preferred
GoldBio s ReadyLadder 100 bp PLUS DNA Ladder is a versatile ready-to-use molecular weight marker designed for sizing small- to mid-range DNA fragments in agarose gel electrophoresis With 12 clearly defined bands spanning 100 bp to 3 000 bp this ladder is well-suited for applications that require precision across a broader range of low molecular weight DNA
Catalog No. 50-005-62121 Supplier Gold Biotechnology Inc Supplier No. D0033000
GoldBio s ReadyLadder 100 bp PLUS DNA Ladder is a versatile ready-to-use molecular weight marker designed for sizing small- to mid-range DNA fragments in agarose gel electrophoresis With 12 clearly defined bands spanning 100 bp to 3 000 bp this ladder is well-suited for applications that require precision across a broader range of low molecular weight DNA