Learn More
Gold Biotechnology Inc ReadyLadderTM 100bp DNA Ladder
SDP
Catalog No. 5000562084
Encompass_Preferred
GoldBio s ReadyLadder 100 bp DNA Ladder is a convenient ready-to-use molecular weight marker designed for accurate sizing of small DNA fragments Composed of 11 defined bands ranging from 100 bp to 1 500 bp this ladder is ideal for applications requiring high resolution of low molecular weight DNA
Catalog No. 50-005-62084 Supplier Gold Biotechnology Inc Supplier No. D0011500
GoldBio s ReadyLadder 100 bp DNA Ladder is a convenient ready-to-use molecular weight marker designed for accurate sizing of small DNA fragments Composed of 11 defined bands ranging from 100 bp to 1 500 bp this ladder is ideal for applications requiring high resolution of low molecular weight DNA