Learn More
Omega Engineering, Inc. Ready-Made Insulated Thermocouples with Kapton, PFA, Glass Braid Insulation and Molded Connectors, 5/pk
Catalog No. NC1651408
Encompass
Made from Special Limits of Error Wire. 1 and 2 meter (36 and 72") Lengths Standard. Molded Subminiature Connector with Integral Strain Relief/Spool.
Catalog No. NC1651408 Supplier Omega Engineering, Inc. Supplier No. 5SRTCTTT3072
Made from Special Limits of Error Wire. 1 and 2 meter (36 and 72") Lengths Standard. Molded Subminiature Connector with Integral Strain Relief/Spool.