Learn More
Sigma Aldrich Fine Chemicals Biosciences ReadyMixTM Taq PCR Reactio
Catalog No. 502900676
Encompass
ReadyMix(TM) Taq PCR Reaction Mix is a prepared SOLUTIONon containing everything needed for a PCR reaction except the specific primers and template. The mix includes Sigmas high quality Taq DNA Polymerase 99% pure deoxynucleotides and buffer in a 2x optimized reaction concentrate. for reaction set-up add the ReadyMix (25 muL) to the primers template and water (total volume 50 muL). Using ReadyMix Taq PCR Reaction Mix reduces pipetting steps and risk of contamination. This saves time and reduces errors while still providing the great performance of Sigmas Taq DNA Polymerase.
Catalog No. 50-290-0676 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P4600100RXN
ReadyMix(TM) Taq PCR Reaction Mix is a prepared SOLUTIONon containing everything needed for a PCR reaction except the specific primers and template. The mix includes Sigmas high quality Taq DNA Polymerase 99% pure deoxynucleotides and buffer in a 2x optimized reaction concentrate. for reaction set-up add the ReadyMix (25 muL) to the primers template and water (total volume 50 muL). Using ReadyMix Taq PCR Reaction Mix reduces pipetting steps and risk of contamination. This saves time and reduces errors while still providing the great performance of Sigmas Taq DNA Polymerase.