Learn More
Bio Rad Laboratories READYPREP 2-D STARTER KIT
Catalog No. 509010217
Encompass
Kit, Starter; Bio-Rad; ReadyPrep 2-D; Includes protein sample, reagents to rehydrate, focus, transfer to second-dimension gels 6 17cm, 10 11cm, or 16 7cm ReadyStrip IPG strips (Kit excludes IPG strips, precast SDS-PAGE gels, gel stains)
Catalog No. 50-901-0217 Supplier Bio Rad Laboratories Supplier No. 1632105
Kit, Starter; Bio-Rad; ReadyPrep 2-D; Includes protein sample, reagents to rehydrate, focus, transfer to second-dimension gels 6 17cm, 10 11cm, or 16 7cm ReadyStrip IPG strips (Kit excludes IPG strips, precast SDS-PAGE gels, gel stains)