Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ReadyProbes™ 2.5% Normal Horse Serum (1X)

Catalog No. R37625
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37625 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37625 Supplier Invitrogen™ Supplier No. R37625
  • $175.00
    $143.50 / Each of 1

    Save $31.50

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments

  • ReadyProbes 2.5% Normal Horse Serum is a ready-to-use non-specific blocker for use in immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF) experiments, usually when a horse or donkey secondary antibody is used.
  • Incubating cells/tissue with diluted serum from the same or similar species as the host of the secondary antibody typically reduces non-specific binding.
  • Simply cover the tissue section or cells with a few drops of ReadyProbes 2.5% Normal Horse Serum and incubate for 30–60 minutes at room temperature.
  • Ready Probes 2.5% Normal Horse Serum can also be used to dilute primary and secondary antibodies.
  • Ready Probes 2.5% Normal Horse Serum is provided as a ready-to-use reagent with no further dilution or preparation required.
  • The serum is collected from healthy adult animals and the heat-treated, centrifuged, and filtered. It is then checked for cross reactivity using an appropriate immunoassay.
  • The serum is diluted to 2.5% (v/v) in buffer, with 0.08% sodium azide added as a preservative.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Provided as 50-mL solution at ready-to-use (1X) concentration in a clear plastic dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Species Horse
Form Liquid
Product Type Horse Serum
Quantity 50 mL
Product Line ReadyProbes
Reagent Type Blocking Reagent
Shipping Condition Room Temperature or Wet Ice
What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Can I use ReadyProbes 2.5% Normal Horse Serum (1X) as an antibody diluent for both primary and secondary antibodies?

Yes, ReadyProbes 2.5% Normal Horse Serum (1X) can be used as an antibody diluent for both primary and secondary antibodies, except for HRP-conjugated antibodies. The sodium azide added as a preservative is an inhibitor of horseradish peroxidase.

How many drops of ReadyProbes 2.5% Normal Horse Serum (1X) should I add to a sample?

Add a sufficient number of drops of ReadyProbes 2.5% Normal Horse Serum (1X) to completely cover the sample.

When would I use goat, horse, or chicken serum for blocking versus FBS or BSA?

For blocking, you should use a serum of the same or closely related species of the secondary antibody being used. If using a secondary antibody that was raised in goat or sheep, use goat serum (Cat. No. R37624 or 31873) for blocking; if using a secondary antibody raised in donkey or horse, use horse serum (Cat. No. R37625 or 31874) for blocking; if using a secondary antibody raised in chicken, use chicken serum (Cat. No. R37626) for blocking. FBS and BSA are alternatives when a specific species of serum is not available.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.