Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ReadyProbes™ Endogenous HRP and AP Blocking Solution (1X)

Catalog No. R37629
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37629 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37629 Supplier Invitrogen™ Supplier No. R37629
  • $221.00
    $181.22 / Each of 1

    Save $39.78

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissue sections, frozen tissue sections, and cell preparations

  • ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activity in formalin-fixed paraffin-embedded tissue sections, frozen tissue sections, and cell preparations.
  • It is provided in a ready-to-use format—just add a few drops and incubate for 10–15 minutes.
  • Endogenous peroxidase, pseudoperoxidase, and alkaline phosphatase activities become problematic when alkaline phosphatase (AP)- or horseradish peroxidase (HRP)- based detection systems are used.
  • ReadyProbes Endogenous Peroxidase and Alkaline Phosphatase Blocking Solution inhibits these endogenous activities, for reduced non-specific signal in immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence (IF), and in situ hybridization (ISH) procedures. This blocker is used before any standard, non-specific blocking solutions (e.g., diluted serum).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Blocking Solution
Quantity 100 mL
Product Line ReadyProbes
Form Liquid
Content And Storage Provided as 100-mL solution at ready-to-use (1X) concentration in a white dropper bottle.

Store at 2–8°C.

When stored as directed, product can be used for 6 months after receipt.

Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
What is the volume for one drop from the ReadyProbes dropper bottles?

The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on cell or tissue lysates?

We do not recommend using ReadyProbes Endogenous HRP and AP Blocking Solution (1X) on cell or tissue lysates. The ReadyProbes Endogenous HRP and AP Blocking Solution (1X) must be removed prior to the application of any HRP- or AP-conjugated antibody/streptavidin. If analyzing the lysates directly, the ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be removed from the lysate.

Can ReadyProbes Endogenous HRP and AP Blocking Solution (1X) be used on live cells?

ReadyProbes Endogenous HRP and AP Blocking Solution (1X) cannot be used on live cells.

How does ReadyProbes Endogenous HRP and AP Blocking Solution (1X) work?

ReadyProbes Endogenous HRP and AP Blocking Solution (1X) works by irreversibly inactivating the enzymes.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.