Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ReadyProbes™ Tissue Autofluorescence Quenching Kit

Catalog No. R37630
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R37630 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R37630 Supplier Invitrogen™ Supplier No. R37630
  • $257.00
    $210.74 / Each of 1

    Save $46.26

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin

  • The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes unwanted autofluorescence in FFPE and frozen tissues due to aldehyde fixation, red blood cells, and structural elements such as collagen and elastin.
  • If the target signal is low enough, then this background signal can interfere with the interpretation of immunohistochemistry (IHC), immunocytochemistry (ICC), immunofluorescence(IF), and in situ hybridization (ISH) assays.
  • The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes most kinds of autofluorescence, with the exception of autofluorescence from lipofuschin granules.
  • The ReadyProbes Tissue Autofluorescence Quenching Kit contains three components that are combined just before use. Combine components A, B, and C in a 1:1:1 ratio.
  • Mix components A and B together before adding component C. The resulting solution is then applied to the tissue for 2–5 minutes at room temperature.
  • This is done after the blocking, primary antibody, and secondary antibody steps, but before application of any counterstains like nuclear stains.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit
Content And Storage A kit containing 5 mL each ready-to-use (1X) solutions of reagent A, B, and C in three clear HDPE bottles.
  • Store at 2–8°C.
  • When stored as directed, product can be used for 3 months after receipt.One kit is sufficient to tre
    Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
    Product Line ReadyProbes
    Product Type Blocking Reagent
    What is the volume for one drop from the ReadyProbes dropper bottles?

    The volume for one drop from the ReadyProbes dropper bottles is approximately 41 µL.

    Does the ReadyProbes Tissue Autofluorescence Quenching Kit quench all autofluorescence?

    No. The ReadyProbes Tissue Autofluorescence Quenching Kit minimizes autofluorescence created by aldehyde fixation, red blood cell autofluorescence, and autofluorescence of collagen and elastin. It will not quench natural pigments or other autofluorescent entities.

    Will the ReadyProbes Tissue Autofluorescence Quenching Kit still work if I messed up the order of addition, i.e., if I mixed Component A and Component C (instead of Component B) and then added Component B?

    For the ReadyProbes Tissue Autofluorescence Quenching Kit to work, the components need to be mixed in the recommended order. Component A needs to be mixed with Component B, followed by mixing in Component C.

    With the ReadyProbes Tissue Autofluorescence Quenching Kit, once I mix Components A, B, and C together, can I store this mixture in the refrigerator or freezer for later use?

    Once the 3 components are mixed together, we recommend using this mixture within 1 hour. The mixture should not be stored beyond 1 hour.

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.